Thermo Scientific™ Nalgene™ Polysulfone Reusable Bottle Top Filters
Convenient filling and vacuum filtration without holding filter to bottle
$156.48 - $156.48
Products ${productFamilyLength}
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
09-740-49C Thermo Scientific™
N7103200045
Case of 5 Each for $156.48
Specifications
Description
  • Available in 250 and 500mL sizes
  • Polysulfone is strong, washable and autoclavable
  • It is noncytotoxic
  • Molded-in graduations make it easy to see sample volume
  • Polypropylene tubing adapter can be stuffed with cotton for sterile venting during autoclaving
  • Removable sterilization membrane support plate is designed to provide maximum flow rate and throughput
  • Accepts 0.25 to 0.3125 in. (6 to 8mm) I.D. vacuum tubing
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.