Thermo Scientific™ Modular Block Accessories
Accessories for modular block dry baths
$194.69 - $194.69
Products 1
Catalog Number Mfr. No. Holds Price Quantity  
Catalog Number Mfr. No. Holds Price Quantity  
22-312-667
View Documents
Thermo Scientific™
2064Q*
1-well modular block
Each for $194.69
Description
Description
  • For use with the digital 2000 series and analog 2050 series modular block dry baths
  • Designed to provide close contact of tubes to block walls for excellent heat transfer
  • Block W x H x D: 9.5 x 5.1 x 7.4cm (3.75 x 2 x 2.9 in.)
SDS