missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Matrix™ Standard Tips

$313.46 - $381.49

Specifications

For Use With (Equipment) Matrix™ electronic pipette
View More Specs

Products 3
Catalog Number Mfr. No. Volume (Metric) Sterility Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Sterility Price Quantity  
14-387-090 View Documents Thermo Scientific™
MX7311
300μL Nonsterile Case of 300 Each for $313.46
 
14-387-091 View Documents Thermo Scientific™
MX7312
300μL Sterile Case for $346.51
 
14-387-098 View Documents Thermo Scientific™
MX8601
625μL Nonsterile Case of 625 Each for $381.49
 
Specifications

Specifications

Matrix™ electronic pipette