missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Low Protein Binding Collection Tubes, 2.0 mL

Minimize protein sample loss during sample manipulation, especially when there are many processing steps, by using Thermo Scientific™ Pierce™ Low Protein Binding Microcentrifuge Tubes (2.0 mL).

$35.35 - $236.00

Specifications

Certifications/Compliance RNase-free, DNase-free
Closure Material Polypropylene
For Use With (Application) Protein biology, protein purification and isolation, protein purification, affinity purification, protein mass spectrometry analysis, sample prep for mass spectrometry
Format Tubes
Formulation Clear, colorless polypropylene microcentrifuge tubes
View More Specs

Products 2
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
PI88379 View Documents Thermo Scientific™
88379
Pack of 250 Each for $35.35
PI88380 View Documents Thermo Scientific™
88380
Case of 2500 Each for $236.00
Description

Description

Thermo Scientific Pierce Low Protein Binding Microcentrifuge Tubes (2.0mL) help minimize protein sample loss during sample manipulation and are ideal in applications where processing steps are extensive.

  • Durable - able to tolerate centrifuge speeds up to 20,000 RCF
  • Compatible - suitable for IP and mass spectrometry applications
  • Nuclease-free - certified RNase/DNase-free
The Pierce Low Protein Binding Microcentrifuge Tubes help minimize sample loss in mass spectrometry and immunoprecipitation/co-immunoprecipitation applications. The polypropylene tubes are certified as DNase/RNase-free and are conveniently packaged (250 tubes per package) to minimize contamination.

Specifications

Specifications

RNase-free, DNase-free
Protein biology, protein purification and isolation, protein purification, affinity purification, protein mass spectrometry analysis, sample prep for mass spectrometry
Clear, colorless polypropylene microcentrifuge tubes
2.0mL
Polypropylene
Low Protein Binding Collection Tubes, 2.0 mL
Polypropylene
Tubes
20,000rcf
2mL
20,000rcf