missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Low Profile Strip Tubes & Caps

Low profile strip tubes for PCR and qPCR applications.

$400.24 - $507.67

Specifications

Certifications/Compliance Certified free from human DNA, RNases, and DNases
View More Specs

Products 6
Catalog Number Mfr. No. Color Closure Included Price Quantity  
Catalog Number Mfr. No. Color Closure Included Price Quantity  
AB0776 View Documents Thermo Scientific™
AB0776
Clear Flat Caps Pack of 250 Each for $425.73
 
AB0775 View Documents Thermo Scientific™
AB0775
Clear Dome Caps Pack of 250 Each for $424.03
 
AB0777 View Documents Thermo Scientific™
AB0777
Assorted Dome Caps Pack of 250 Each for $507.67
 
AB0778 View Documents Thermo Scientific™
AB0778
Assorted Flat Caps Pack of 250 Each for $481.97
 
AB1770 View Documents Thermo Scientific™
AB1770
Clear Ultra-clear Dome Caps Pack of 250 Each for $400.24
 
AB1771 View Documents Thermo Scientific™
AB1771
White Ultra-clear Dome Caps Pack of 250 Each for $456.20
 
Description

Description

The low profile design of the Thermo Scientific Low Profile Strip Tubes reduces the dead space between the heated lid of the thermal cycler and the sample. This eliminates condensation on the side wall of the tube, preventing reduction in PCR volume and increasing the efficiency of the reaction. Thermo Scientific low profile products are especially recommended for use with reaction volumes below 20 μL.

The low profile design of the Thermo Scientific strip tubes without caps reduces the 'dead space' between the heated lid of the thermal cycler and the sample. This eliminates condensation on the side wall of the tube, preventing reduction in PCR volume and increasing the efficiency of the reaction. Low profile products are especially recommended for use with reaction volumes below 20 μL.

Highlights

  • Suitable for 0.2 mL thermal cycler blocks
  • 150 μL maximum tube capacity when capped
  • Low profile to reduce dead space and increase PCR efficiency
  • Labelled A-H end tabs (8-strip) and 1-12 end tabs (12-strip)
Applications
  • PCR
  • qPCR/Real-Time PCR

Hazardous: No
Storage Condition: Ambient

PCR, qPCR and Real-Time PCR.

Certifications

Certified free from human DNA, RNases, and DNases.

Specifications

Specifications

Certified free from human DNA, RNases, and DNases