Thermo Scientific™ Cell Roller System Accessories for Environmental Chambers and Incubators
Allow production of monolayer cell cultures in standard roller culture vessels.
Manufacturer:  Thermo Scientific™ FOR500182
 View more versions of this product
Catalog No. 15-465-185
Description
Description
Allow production of monolayer cell cultures in standard roller culture vessels with these Cell Roller System Accessories for Thermo Scientific™ Environmental Chambers and Incubators. Oxygenation and exposure of the cells to the media growth area are improved. Culture yields are increased by the uniform temperature control and cell roll system’s continuous, gentle rotation.
Specifications
Specifications
Accommodates a cell roller
Customer installed
Reinforced Floor Options
Reinforced Floor with Removable Ramp
Model 3948 and 3962 Environmental Chamber/Incubator
Documents