Thermo Scientific™ Jewett™ General Purpose Refrigerator/Freezer Combinations
Designed for dependable, energy-efficient performance
$6,871.37 - $6,871.37
Includes: Power cord and plug
Products ${productFamilyLength}
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
22-650-917 Thermo Scientific™
R2501B
Each for $6,871.37
Specifications
Description
  • Operating temperature ranges: 2° to 10°C (refrigerator), -12° to -18°C (freezer)
  • White acrylic exterior and seamless interior finishes
  • Steel-encased electrical components
  • Spark-proof interiors with nonsparking magnetic door gasket
  • Foamed-in-place, CFC-free urethane insulation
  • Hermetically sealed compressor
Warranty
2 years parts, 1 year labor, 3 years additional compressor parts only; 16 months parts only (International)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.