Please login to your online account to display your discounted pricing
Thermo Scientific™ Jewett™ General-Purpose Freezers
Designed for dependable, energy-efficient performance
$3,219.85 - $3,219.85
Specifications
Hertz 60Hz
Temperature Range (Metric) −12° to −20°C
View More Specs
Products
Catalog Number Mfr. No. Voltage Price Quantity    
22-650-899
thermo scientific™
C65F1B
120V Each for $3,219.85
Description & Specifications
Specifications
Hertz 60Hz
Temperature Range (Metric) −12° to −20°C
Our Jewett general-purpose freezers provide cost-effective, reliable storage. Refrigeration systems are equipped with reliable, hermetically sealed compressors for quiet operation.
  • Steel-encased electrical components
  • Spark-proof interiors with nonsparking magnetic door gasket
  • White acrylic exterior and seamless interior finishes
  • Foamed-in-place, CFC-free urethane insulation
  • Hermetically sealed compressor
UL listed