Please login to your online account to display your discounted pricing
Thermo Scientific™ IEC™ Multi™ and MultiRF™ Four-Place Swinging Bucket Rotor::Aerocarrier™ Adapters
$689.30 - $802.25
Products
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity    
04-976-015
thermo scientific™
06556EF
Thermo Scientific™ IEC™ Multi and MultiRF swing bucket rotor for CL3/CL3R benchtop centrifuge Pack of 2 Each for $738.57
27-021-541
thermo scientific™
06558EF
Thermo Scientific™ IEC™ Multi and MultiRF swing bucket rotor for CL3/CL3R benchtop centrifuge Pack of 2 Each for $701.40
04-976-018
thermo scientific™
06559EF
Thermo Scientific™ IEC™ Multi and MultiRF swing bucket rotor for CL3/CL3R benchtop centrifuge Pack of 2 Each for $733.11
05-375-51
thermo scientific™
06560EF
05-375-76 (in Multi/MultiRF) and 05-375-45 (in Centra-CL3/CL3R) Pack of 2 Each for $701.40
05-375-52
thermo scientific™
06561EF
Thermo Scientific™ IEC™ Multi and MultiRF swing bucket rotor for CL3/CL3R benchtop centrifuge Pack of 2 Each for $696.21
05-375-53
thermo scientific™
06562EF*
Thermo Scientific™ IEC™ Multi and MultiRF swing bucket rotor for CL3/CL3R benchtop centrifuge Pack of 2 Each for $689.30
05-375-54
thermo scientific™
06563EF
Thermo Scientific™ IEC™ Multi and MultiRF swing bucket rotor for CL3/CL3R benchtop centrifuge Pack of 2 Each for $738.57
05-375-56
thermo scientific™
06565EF
Thermo Scientific™ IEC™ Multi and MultiRF swing bucket rotor for CL3/CL3R benchtop centrifuge Pack of 2 Each for $701.40
05-375-57
thermo scientific™
06566EF
Thermo Scientific™ IEC™ Multi and MultiRF swing bucket rotor for CL3/CL3R benchtop centrifuge Pack of 2 Each for $689.30
05-375-58
thermo scientific™
06567EF
Thermo Scientific™ IEC™ Multi and MultiRF swing bucket rotor for CL3/CL3R benchtop centrifuge Pack of 2 Each for $802.25
05-375-59
thermo scientific™
06568EF
Thermo Scientific™ IEC™ Multi and MultiRF swing bucket rotor for CL3/CL3R benchtop centrifuge Pack of 2 Each for $729.04
Description & Specifications
For use in 05-375-76 (with Multi/MultiRF) and 05-375-45 (with Centra-CL3/-CL3R) swinging-bucket rotors.
  • With threaded transparent sealing cover with O-ring to provide aerosol containment when running toxic, infectious, or sterile samples
  • Polypropylene, can be cleaned with bleach or decontamination washes
Carriers are sold in packs of two; purchase the proper number to fully equip rotor.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.