missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Hypersil™ Silica HPLC Columns, 5μm Particle Size

An unbonded media providing an efficient tool for normal phase chromatography of nonpolar and moderately polar organic compounds

Manufacturer:  Thermo Scientific™ 30005052130

 View more versions of this product

Catalog No. 30005052130



Description

Description

  • Excellent batch-to-batch reproducibility
  • Spherical particle with narrow particle size distribution
  • Long column lifetime
  • High performance and column efficiency

Order Info

Other column dimensions are available. Also available in 10μm. Please call Customer Service for more information.

Specifications

Specifications

Normal Phase
Spherical, Fully Porous Type A Silica
Normal Phase
2.1 mm
5800 psi (400 bar)
2 to 8
170 m²/g
L3
No
0%
Analytical Column
50 mm
5 μm
120 Å
60°C
Silica
SDS
Documents