Please login to your online account to display your discounted pricing
Thermo Scientific™ Hypersil™ MOS Drop-in Guard Cartridges
Offer convenience, economy and effective protection for extending column lifetime
$503.28 - $503.28
Specifications
Length (Metric) 10mm
Stationary Phase C8
View More Specs
Products
Catalog Number Mfr. No. Diameter (Metric) Inner Particle Size Price Quantity    
03-150-543
thermo scientific™
30205012101
2.1mm 5μm Pack of 4 Each for $503.28
03-150-544
thermo scientific™
30205013001
3mm 5μm Pack of 4 Each for $503.28
Description & Specifications
Specifications
Length (Metric) 10mm
Stationary Phase C8
  • The 10mm design offers maximum protection with minimal increase in retention
  • Fit Thermo Scientific UNIGUARD direct connection and stand alone holders
  • Available in packs of 4
Feature of MOS (C8) Solid Phase
  • Reliable, less retentive phase