Thermo Scientific™ Hypersil™ MOS-2 (C8) Columns, 3μm Particle Size
Have a monolayer coverage of C8 alkyl chain chemically bonded onto the silica surface for a reproducible and efficient stationary phase
Manufacturer:  Thermo Scientific™ 30303104630
 View more versions of this product
Catalog No. 03-050-500
Description
Description
  • Reliable, less retentive phase
  • Long column lifetimes, even under basic conditions
  • Manufactured under ISO 9001-2000 quality guidelines
  • Affordable
Specifications
Specifications
4.6 mm
100 mm
3 µm
MOS-2 (C8)
Documents