Please login to your online account to display your discounted pricing

Thermo Scientific™ Hypersil™ MOS-2 (C8) Columns, 3μm Particle Size

Have a monolayer coverage of C8 alkyl chain chemically bonded onto the silica surface for a reproducible and efficient stationary phase

$1,067.08 - $1,085.65

Specifications

Diameter (Metric) Inner 4.6mm
Particle Size 3μm
Stationary Phase MOS-2 (C8)
View More Specs
Products
Catalog Number Mfr. No. Length (Metric) Price Quantity    

03-050-500

 
thermo scientific™
30303104630
100mm Each for $1,085.65

30303-154630

 
thermo scientific™
30303154630
150mm Each for $1,067.08
Description & Specifications

Specifications

Diameter (Metric) Inner 4.6mm
Particle Size 3μm
Stationary Phase MOS-2 (C8)

  • Reliable, less retentive phase
  • Long column lifetimes, even under basic conditions
  • Manufactured under ISO 9001-2000 quality guidelines
  • Affordable