Please login to your online account to display your discounted pricing

Thermo Scientific™ Hypersil™ GOLD HPLC Columns, 3μm Particle Size

Endcapped, ultrapure, silica-based columns with exceptional peak shape and resolution for HPLC and LC/MS

$1,329.94 - $1,338.25

Specifications

Particle Size 3μm
View More Specs
Products
Catalog Number Mfr. No. Diameter (Metric) Inner Stationary Phase Length (Metric) Price Quantity    

03-260-110

 
thermo scientific™
25003254030
4mm C18 250mm Each for $1,338.25

25003022130

 
thermo scientific™
25003022130
2.1mm C18 20mm Each for $1,329.94

25003023030

 
thermo fisher scientific
25003023030
3mm C18 20mm Each for $1,329.94
Description & Specifications

Specifications

Particle Size 3μm

  • Significant reduction in peak tailing while retaining C18 selectivity
  • Excellent resolution, efficiency and sensitvity
  • Confidence in the accuracy and quality of analytical data

Other custom column dimensions are available. Please call your local Customer Service for more information.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.