Please login to your online account to display your discounted pricing
Thermo Scientific™ Hypersil™ GOLD CN Preparative HPLC Columns, 5μm Particle Size
Preparative columns are available in popular Thermo Scientific phases
$2,182.69 - $8,072.95
Specifications
Particle Size 5μm
Stationary Phase CN
View More Specs
Products
Catalog Number Mfr. No. Diameter (Metric) Inner Length (Metric) Price Quantity    
03-051-138
thermo scientific™
25805109070
10mm 100mm Each for $2,546.82
03-051-134
thermo scientific™
25805059270
21mm 50mm Each for $4,441.83
03-051-133
thermo scientific™
25805059070
10mm 50mm Each for $2,182.69
03-051-135
thermo scientific™
25805059370
30mm 50mm Each for $8,072.95
03-051-136
thermo scientific™
25805059570
50mm 50mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
03-051-140
thermo scientific™
25805109370
30mm 100mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
03-051-141
thermo scientific™
25805109570
50mm 100mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
03-051-144
thermo scientific™
25805159270
21mm 150mm Each for $5,762.54
03-051-145
thermo scientific™
25805159370
30mm 150mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
03-051-146
thermo scientific™
25805159570
50mm 150mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
03-051-149
thermo scientific™
25805259270
21mm 250mm Each for $7,604.57
03-051-150
thermo scientific™
25805259370
30mm 250mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
03-051-151
thermo scientific™
25805259570
50mm 250mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications
Specifications
Particle Size 5μm
Stationary Phase CN
  • Hypersil GOLD CN columns provide alternative selectivity with lower hydrophobicity
  • Excellent peak shape
  • Outstanding senstivity
  • Less retention for faster analysis
Other custom column dimensions are available. Please call your local Customer Service for more information.