Thermo Scientific™ Hypersil™ GOLD C8 KAPPA Capillary Columns
Narrow ID columns operate at low flow rates required for greater ionization efficiencies
$1,380.43 - $1,477.69
Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (Metric) Length (Metric) Particle Size Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Length (Metric) Particle Size Price Quantity  
25-202-100365 Thermo Scientific™
25202100365
0.32mm 100mm 1.9μm Each for $1,477.69
25-203-100365 Thermo Scientific™
25203100365
0.32mm 100mm 3μm Each for $1,380.43
25-203-100565 Thermo Scientific™
25203100565
0.5mm 100mm 3μm Each for $1,418.40
Specifications
Description
Features of Hypersil GOLD C8 Solid Phase
  • Hypersil GOLD C8 columns provide similar selectivity to C18 columns but with reduced retention
  • Lower hydrophobicity
  • Faster separations
  • Excellent peak shape
  • High efficiency
  • Outstanding sensitivity