Please login to your online account to display your discounted pricing
Thermo Scientific™ Hypersil GOLD™ aQ Nanobore HPLC Columns, 1.9um Particle Size
At flow rates of nL/min versus mL/min, offer greater signal to noise ratio than traditional columns used with electrospray
$803.22 - $803.22
Products
Catalog Number Mfr. No. Price Quantity    
03-051-209
thermo scientific™
25302057581
Each for $803.22
Description & Specifications
Features of Hypersil GOLD aQ Solid Phase
  • Hypersil GOLD aQ columns provide enhanced retention and resolution of polar analytes
  • Polar endcapped C18 phase for alternative selectivity
  • Retention and resolution of polar analytes
  • Excellent for highly aqueous mobile phase
  • Excellent peak shape