Please login to your online account to display your discounted pricing
Thermo Scientific™ Hypersil GOLD™ aQ KAPPA Capillary HPLC Columns, 1.9μm Particle Size
Narrow ID columns operate at low flow rates required for greater ionization efficiencies
$1,518.32 - $1,518.32
Specifications
Diameter (Metric) Inner 320 Glc muM
Particle Size 1.9μm
Stationary Phase aQ, C18
View More Specs
Products
Catalog Number Mfr. No. Length (Metric) Price Quantity    
03-051-211
thermo scientific™
25302100365
100mm Each for $1,518.32
Description & Specifications
Specifications
Diameter (Metric) Inner 320 Glc muM
Particle Size 1.9μm
Stationary Phase aQ, C18
High efficiency KAPPA columns offer high performance and sensitivity.
  • 75μm to 500μm ID
  • Rugged sleeved design for robustness
  • Ideal for LC/MS analysis
  • Hypersil GOLD aQ columns provide enhanced retention and resolution of polar analytes.
  • Polar endcapped C18 phase for alternative selectivity
  • Retention and resolution of polar analytes
  • Ideal for highly aqueous mobile phase
  • Excellent peak shape
Other custom column dimensions are available. Please call your local Customer Service for more information.