Thermo Scientific™ Hypersil GOLD™ aQ KAPPA Capillary HPLC Columns, 1.9μm Particle Size
Narrow ID columns operate at low flow rates required for greater ionization efficiencies
Manufacturer:  Thermo Scientific™ 25302100365
 View more versions of this product
Catalog No. 03-051-211
Description
Description
High efficiency KAPPA columns offer high performance and sensitivity.
  • 75μm to 500μm ID
  • Rugged sleeved design for robustness
  • Ideal for LC/MS analysis
  • Hypersil GOLD aQ columns provide enhanced retention and resolution of polar analytes.
  • Polar endcapped C18 phase for alternative selectivity
  • Retention and resolution of polar analytes
  • Ideal for highly aqueous mobile phase
  • Excellent peak shape
Order Info
Other custom column dimensions are available. Please call your local Customer Service for more information.
Specifications
Specifications
320 Glc muM
100 mm
1.9 µm
aQ, C18
Documents