Please login to your online account to display your discounted pricing
Thermo Scientific™ Hypersil GOLD™ aQ Javelin™ Guard HPLC Columns
Javelin guard columns offer convenient, easy to use protection for analytical HPLC columns
$828.46 - $828.46
Specifications
Length (Metric) 10mm
Particle Size 5μm
Stationary Phase aQ
View More Specs
Products
Catalog Number Mfr. No. Diameter (Metric) Inner Price Quantity    
03-051-246
thermo scientific™
25305012106
2.1mm Pack of 4 Each for $828.46
Description & Specifications
Specifications
Length (Metric) 10mm
Particle Size 5μm
Stationary Phase aQ
  • Direct-connection, patented fingertight design has a 1/16 in. CPI tip that attaches directly to the inlet of the HPLC column with tubing or wrenches
  • Requires no holder, allowing quick and easy replacement
Hypersil GOLD aQ columns provide enhanced retention and resolution of polar analytes.
  • Polar endcapped C18 phase for alternative selectivity
  • Retention and resolution of polar analytes
  • Ideal for highly aqueous mobile phase
  • Excellent peak shape
Javelin guards are available in other Thermo Scientific phases. Please call your local Customer Service for more information.