Please login to your online account to display your discounted pricing
Thermo Scientific™ Hypercarb™ Drop-in Guard Cartridges
Offer convenience, economy and effective protection for extending column lifetime
$534.91 - $534.91
Specifications
Length (Metric) 10mm
View More Specs
Products
Catalog Number Mfr. No. Particle Size Price Quantity    
03-170-491
thermo scientific™
35007014001
7μm Pack of 2 Each for $534.91
Description & Specifications
Specifications
Length (Metric) 10mm
  • The 10mm design offers maximum protection with minimal increase in retention
  • Compatible with Thermo Scientific Uniguard™ direct connection and stand-alone holders
Features of Hypercarb Solid Phase
  • Suited to solve problem separations
  • Exceptional retention of very polar analytes
  • True orthoganality to C18
  • Separate structurally related substances
  • Excellent for high temperature applications
Drop-in cartridges are available in other Thermo Scientific phases. Please contact your Customer Service Representative for more information