missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Epredia™ HM 650V Vibrating Blade Microtome Accessories
Supplier: Epredia™ 419150
Description
Standard buffer tray
- Holds stages in place using a magnet
- Fill tray with all water-based buffers and solutions
- Use pre-cooled in the standard buffer tray for cooling tissue
- Use frozen together with buffer solution to prevent the frozen buffer solution from inflating
- Magnification 2.5X
- Move, swivel, and displace for optimal viewing position
- Includes adapter and illumination
- Eyepieces: 10X
- Supplementary lens: 0.63X
- Zoom magnifying range: 4.1 to 31.5X
- Microscope adapter for angled view of specimen
- Horizontal view and vertical drive and swivel mechanism
- 3.5mm fiber bundle
- Swan-neck light guide: 50mm long
- Flexible light guide: 750mm long
- High light intensity
- Include adapter for the KL200 (760317)
Specifications
Microtome Blade Holder |