Please login to your online account to display your discounted pricing
Thermo Scientific™ Heracell™ VIOS 250i CO2 Incubator O2 Control Glass Doors
O2 control glass doors for VIOS 250i CO2 incubators designed for sensitive cultures like stem and primary cells in leading research, pharmaceutical and clinical laboratory applications
$8,364.49 - $8,364.49
For Use With (Equipment) Heracell VIOS 250i CO2 Incubator
View More Specs
Products
Catalog Number Mfr. No. Item Description Price Quantity    
13998161
thermo scientific™
51901134
O2 5% - 90% Control Glass Doors Each for $8,364.49
13998160
thermo scientific™
51901133
O2 1-21% Control Glass Doors Each for $8,364.49
Description & Specifications
Specifications
For Use With (Equipment) Heracell VIOS 250i CO2 Incubator
Includes O2 control with gas tight screen, 6 inner glass doors,1/2 width shelves
Material Glass
  • Gas tight screen
  • 6 inner glass doors
  • 1/2 width shelves