Thermo Scientific™ Heracell™ External Guard for CO2 Incubator Gas Regulators
N2 and O2 for Heracell CO2 incubators
$2,273.98 - $2,273.98
Specifications
Products ${productFamilyLength}
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
13-998-072 Thermo Scientific™
50059043
External Gas Tank Guard Each for $2,273.98
Specifications
Description
  • Two-stage gas tanks for N2 or O2.
  • External gas guard automatic change-over to reserve tank
  • 120 V, 50/60 Hz
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
0.0