Thermo Scientific™ Heracell™ 150i Tri-Gas Incubator Packages
Choice of noncorrosive stainless steel or 100% pure copper
Specifications
Controller Type Touchscreen
Sensor Types TC sensor
Warranty 2 Years
View More Specs
Alert:
Products on this page may include voltages not considered standard in North America. Please review carefully.
Products ${productFamilyLength}
Catalog Number Mfr. No. Chamber Material Voltage Price Quantity  
Catalog Number Mfr. No. Chamber Material Voltage Price Quantity  
11-687-201 Thermo Scientific™
50116054
Copper 120V N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
11-687-200 Thermo Scientific™
50116052
Stainless Steel 120V N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Description
Thermo Scientific Heracell i tri-gas incubators feature innovative ContraCon moist heat decontamination technology that ensures worry-free cleaning and operation. These incubators are designed to provide optimal growth conditions and deliver superior parameter recovery rates.
iCAN™ Touchscreen Control
  • Door-mounted for easy accessibility and viewing
  • Convenient on-screen user prompts for simplified measurement and control navigation
  • Choice of languages
  • Changes in culture environment visible on screen logs and usage recording
  • Visual monitor alarm
TC or IR CO2 Sensor
  • Standard thermal conductivity (TC) sensor features accurate gas sensor technology
  • Infrared (IR) sensor features exclusive split-beam technology for precise CO2 control in fluctuating temperature and humidity conditions
Rapid Humidity Recovery
  • Large, directly heated water reservoir
  • Recovery times are up to five times faster than conventional water pans
  • Water level alarm with audible and visual alerts
Optional O2 Control with Auto-Cal
  • Two O2 control ranges precisely control oxygen concentration
  • Ideal for applications requiring decreased or increased O2 conditions
  • Zirconium Oxide sensor
Warranty
Two years
Description & Specifications
Specifications
Touchscreen
TC sensor
2 Years