missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Heracell™ 150i Tri-Gas Incubator Packages

Choice of noncorrosive stainless steel or 100% pure copper

Manufacturer:  Thermo Scientific™ 50116054

 View more versions of this product

Catalog No. 11-687-201

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Alert:

Products on this page may include voltages not considered standard in North America. Please review carefully.


Specifications Description & Specifications

Specifications

2 Years
Copper
60Hz
Touchscreen
TC sensor
120V
120V 60Hz