Please login to your online account to display your discounted pricing
Thermo Scientific™ Heracell™ 150i and 240i CO2 Incubators with Copper Chambers
Touch screen operating simplicity with exclusive 100% pure copper interior chamber
Specifications
Controller Type Optional 1-21% or 5-90%
Data Outputs RS232
Disinfection Time 90°C/9 hr.
Humidity Delivery integral panless system
Hertz 50/60Hz
View More Specs
Products
Catalog Number Mfr. No. Humidity Range Ports Sensor Types Temperature Range (Metric) Price Quantity    
MPP50116050
thermo scientific™
MPP50116050
Up to 95% 1.6 in. (4.2cm) access, with silicon seal TC sensor Ambient 3 to 55°C CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MPP51026283
thermo scientific™
MPP51026283
Up to 95% 1.6 in. (4.2cm) access, with silicon seal TC sensor Ambient 3 to 55°C CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications
Specifications
Controller Type Optional 1-21% or 5-90%
Data Outputs RS232
Disinfection Time 90°C/9 hr.
Humidity Delivery integral panless system
Hertz 50/60Hz
Range (Concentration) 0 to 20%
Shelf Material Solid copper
Thermo Scientific Heracell i CO2 incubators feature innovative ContraCon moist heat decontamination technology that ensures worry-free cleaning and operation. These incubators are designed to provide optimal growth conditions and deliver superior parameter recovery rates.
iCAN™ Touchscreen Control
  • Door-mounted for easy accessibility and viewing
  • Convenient on-screen user prompts for simplified measurement and control navigation
  • Choice of languages
  • Changes in culture environment visible on screen logs and usage recording
  • Visual monitor alarm
TC or IR CO2 Sensor
  • Standard thermal conductivity (TC) sensor features accurate gas sensor technology
  • Infrared (IR) sensor features exclusive split-beam technology for precise CO2 control in fluctuating temperature and humidity conditions
Rapid Humidity Recovery
  • Large, directly heated water reservoir
  • Recovery times are up to five times faster than conventional water pans
  • Water level alarm with audible and visual alerts
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.