Thermo Scientific™ Adapters for HBB-6 Swinging Bucket Rotor
For use with the Thermo Scientific HBB-6 Swinging Bucket Rotor
Manufacturer:  Thermo Scientific™ 11751
 View more versions of this product
Catalog No. 11-751
Description
Description
  • A range of adapters to accommodate a variety of blood bags varying in sizes.
  • Adapters for up to 12 platelet-rich plasma and buffy coat bags
Specifications
Specifications
Single or Double Blood Bag
Adapter
12 x 450 mL
12