Please login to your online account to display your discounted pricing
Thermo Scientific Handheld Digital Pyrometer Type K Thermocouple Probes
$369.35 - $369.35
Products
Catalog Number Mfr. No. Price Quantity    
13-904-50A
thermo sci measurement
TC405X2
Each for $369.35
Description & Specifications
Type-K probes come with 3 ft. (91cm) flexible coiled extension cable; flat, yellow ANSI-coded subminiature thermocouple connector, and 4 in.L (10cm) plastic handle with a maximum exposure temperature of 105°C (221°F).
General-purpose/Immersion Probe
  • Fast response probe for air (especially in furnaces), liquid, semiliquid and granular materials Bend to any shape; Inconel™ Type-600 sheath
  • Penetration Probe
    • For measurement in soft and semifrozen materials like food, tissue cultures, rubber Stainless-steel Type-304 sheath
  • Air Probe
    • For measuring air or gases in turbulent air heating and cooling units and engine exhaust systems Stainless-steel Type-304 sheath
  • Surface Probe
    • Open end for fast response measurement against solid surfaces like hotplates, molds, semiconductors
    • Spring-loaded tip for solid contact; stainless-steel Type-304 sheath