Please login to your online account to display your discounted pricing

Thermo Scientific™ Golden Gate ATR for Nicolet™ iS5 FT-IR Spectrometer

This versatile infrared sampling accessory is fast, sensitive, and robust

Products
Catalog Number Mfr. No. Price Quantity    

14-387-613

 
thermo scientific™
869145300
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

14-387-612

 
thermo scientific™
069145200
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Analyzes sample types from hard solids to corrosive liquids

  • Used to analyze a range of samples from single particles and fibers to corrosive liquids
  • High temperature bonded into its tungsten carbide mount giving performance and strength to withstand the high pressures
  • Used for almost any sample type in any research or QA environment
  • Little or no sample preparation needed
  • Valuable accessory for the preparation of toxic compounds
  • Ideal for macro sampling
  • 45° single reflection Type IIa diamond
  • Brazed tungsten carbide diamond mount
  • Fully enclosed beam condensing lens system
  • Sapphire anvil for high pressure contact
  • Reproducible pressure control to 80 pounds
  • Thermal, cryogenic, and super critical fluid options available
  • Fully enclosed sample anvil available for dangerous or sensitive samples
Specifications
  • Crystal material: Type IIa diamond
  • Spectral wavelength range: 5000 to 650cm–1 ZnSe optics, 5000 to 400cm–1 KRS–5 optics

Polymer pellets, intractable or opaque solids, single fibers, corrosive liquids, geochemical samples, powders, coated wires, air sensitive samples, hazardous materials/toxic compounds