Please login to your online account to display your discounted pricing

Thermo Scientific™ Genesys™ 10S Vis Spectrophotometers

Traditional, grating-based spectrophotometer to teach the basics of spectroscopy.

$5,985.90 - $5,985.90

Specifications

Accuracy (Photometric) 0.5% or ±0.005A, whichever is greater up to 2A
Certifications/Compliance UL/cUL, CSA
Connections USB
Depth (English) 15.7 in.
Depth (Metric) 40cm
View More Specs
Includes:  Spectrophotometer, six-position cell changer, single cell holder, 100-240V automatic power supply, AC power cord, spare fuses, protective plastic cover, USB memory device and USB cable
Products
Catalog Number Mfr. No. Includes Price Quantity    

S99608

 
thermo scientific™
840-20790
Spectrophotometer, six-position cell changer, single cell holder, 100 to 240V automatic power supply, AC power cord, spare fuses, protective Plastic cover, USB memory device and USB cable Each for $5,985.90
Description & Specifications

Specifications

Accuracy (Photometric) 0.5% or ±0.005A, whichever is greater up to 2A
Certifications/Compliance UL/cUL, CSA
Connections USB
Depth (English) 15.7 in.
Depth (Metric) 40cm
Detection Range 325 to 1,100nm
Detector Type Silicon Photodiode
Lamp Tungsten Halogen
Optical Design Single Beam
Spectral Bandwidth 5nm
Display LCD
Electrical Requirements 100 to 240V 50/60Hz
For Use With (Application) Basic QA/QC, routine analysis, teaching and educational laboratories
Height (English) 9.8 in.
Height (Metric) 25cm
Hertz 50/60Hz
Noise >0.001A at 0A; >0.002A at 2A; peak-to peak at 340nm
Scan Speed 10 to 4200nm/min.
Speed 10 to 4,200nm/min.
Voltage 100 to 240V
Wavelength Accuracy ±1nm
Wavelength Range 325 to 1100nm
Wavelength Repeatability ±0.5nm
Weight (English) 19 lb.
Weight (Metric) 8.6kg
Width (English) 11.8 in.
Width (Metric) 30cm

Students can acquire the data in the lab, save it to a memory device and use a separate computer for analyzing data and writing lab reports.

  • Lightweight instruments are easy to transport and store
  • Precision electronics and a simple, single-beam optical geometry provide accurate results
  • Spectral bandwidth is ideal for most routine quality control measurements
  • Perform simple enzymatic assays, color determination, concentration measurements and more
  • Simple-but-powerful embedded software with an easy-to-use interface