Please login to your online account to display your discounted pricing

Thermo Scientific™ Gene Frame Seals

Gene Frames for in-situ feature a dual strength adhesive system which facilitates the easy removal of the frames after use.

$309.52 - $367.70

Specifications

Applications For in-situ hybridization and amplification
Certifications/Compliance Certified free from DNase, RNase and Human Genomic DNA
Includes 100 rigid coverslips
Quantity 100 Pack
View More Specs
Includes:  100 Rigid coverslips for easy handling and uniform hybridizations
Products
Catalog Number Mfr. No. Volume (Metric) Price Quantity    

AB-0576

 
thermo scientific™
AB0576
25μL Pack of 100 Each for $309.52

AB-0577

 
thermo scientific™
AB0577
65μL Pack of 100 Each for $331.09

AB-0578

 
thermo scientific™
AB0578
125μL Pack of 100 Each for $367.70
Description & Specifications

Specifications

Applications For in-situ hybridization and amplification
Certifications/Compliance Certified free from DNase, RNase and Human Genomic DNA
Includes 100 rigid coverslips
Quantity 100 Pack

Thermo Scientific Gene Frames have adhesives on either side of the frame which are of differing strengths; the side which adheres to the glass has an easy release adhesive whereas the reverse, which sticks to the coverslip, forms a stronger bond. This means that when the coverslip is removed, due to the disparity in the strengths of the different adhesives, the frame is removed simultaneously with no sticky residue being left behind on the slide.
Highlights
• Gas tight sealing prevents reagent loss due to evaporation
• Can withstand temperatures of 97°C
• Designed for use with standard microscope slides
• Frame and coverslip can be removed simultaneously leaving no residue
• Can be autoclaved or sterilized by UV irradiation
• All Gene Frame packs are supplied with 100 coverslips
• Rigid coverslips for ease of handling and uniform hybridizations
• Thickness of Gene Frames: 0.25 mm
Applications
• In-situ PCR

Hazardous:No
>Storage Condition:Ambient

Certified free from DNase, RNase and human genomic DNA.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.