Thermo Scientific™ Four-Place Swinging Bucket Rotor and Aerocarrier Sealed Tube Insert for CL2 Centrifuges
For use in 002360F swinging bucket rotor
Manufacturer:  Thermo Scientific™ 02091SF
 View more versions of this product
Catalog No. 05-112-107
Description
Description
Unique polypropylene inserts that serve as complete, self-contained holders for 5 to 10mL blood tubes, 12mL UriSystem™ tubes, or 15/50mL Falcon™ and Corning™ conical tubes.
Tubes Feature
  • Screw-on cap with O-ring closure for complete aerosol containment Transparent lid lets you check tube for breakage before opening lid If breakage occurs, tube inserts can be autoclaved or decontaminated with diluted household bleach
Compliance
Inserts meet OSHA Bloodborne Pathogen Final Rule
Specifications
Specifications
Thermo Scientific™ IEC™ general purpose centrifuge
50mL
Documents