Thermo Scientific™ Four-Place Swinging Bucket Rotor and Aerocarrier Sealed Tube Insert for CL2 Centrifuges
For use in 002360F swinging bucket rotor
$494.38 - $634.17
Products 3
Catalog Number Mfr. No. For Use With (Equipment) Tube Size Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Tube Size Price Quantity  
05-112-106
View Documents
Thermo Scientific™
02092SF
Thermo Scientific™ IEC™ general purpose centrifuge 10 to 15mL
Pack of 2 Each for $496.08
05-112-104
View Documents
Thermo Scientific™
02093SF
Centra CL2 centrifuge 7 to 10mL
Pack of 2 Each for $494.38
05-101-16
View Documents
Thermo Scientific™
2094SF
Centra CL2 centrifuge 5 to 7mL
Pack of 12 Each for $634.17
Description
Description
Unique polypropylene inserts that serve as complete, self-contained holders for 5 to 10mL blood tubes, 12mL UriSystem™ tubes, or 15/50mL Falcon™ and Corning™ conical tubes.
Tubes Feature
  • Screw-on cap with O-ring closure for complete aerosol containment Transparent lid lets you check tube for breakage before opening lid If breakage occurs, tube inserts can be autoclaved or decontaminated with diluted household bleach
Compliance
Inserts meet OSHA Bloodborne Pathogen Final Rule
SDS