Please login to your online account to display your discounted pricing
Thermo Scientific™ Fluophase™ PFP HPLC Columns
Perfluorinated phases provide unique selectivity
$1,036.88 - $1,036.88
Specifications
Diameter (Metric) Inner 2.1mm
Particle Size 5μm
Stationary Phase PFP
View More Specs
Products
Catalog Number Mfr. No. Length (Metric) Price Quantity    
82705052130
thermo scientific™
82705052130
50mm Each for $1,036.88
Description & Specifications
Specifications
Diameter (Metric) Inner 2.1mm
Particle Size 5μm
Stationary Phase PFP
  • Extra retention and selectivity for halogenenated compounds
  • Excellent for taxane analysis
Other column dimensions are available. Please call your Customer Service Representative for more information.