missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Flexaframe™ Castaloy™ Hook Connector

Locks two Flexframe rods at a 90° angle

Manufacturer:  Fisherbrand™ 00529Q

Catalog No. 1466618Q



Description

Description

 

  • Can't slip
  • Install without disassembling frame
  • Supplied with Flexframe sets
  • Holders work with 13mm diameter rods

 

Specifications

Specifications

Locks two rods at 90° angle, slip proof, install without disassembling frame, comes with flexaframe sets
Hook Connector
SDS
Documents
Product Certifications
Promotions

Promotions