Thermo Scientific™ Finntip™ Pocket Tips
Dispense nanoliter volumes with handheld pipettes
$1,216.73 - $1,216.73
Specifications
Packaging 10 x 96/Rack
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
14-487-87 Thermo Scientific™
94035550
250nL Case of 960 Each for $1,216.73
Specifications
Description
Tips have a molded pocket inside to allow transfer of compounds suspended in DMSO into an assay.
  • 50 and 250nl dispensing
  • Works with DMSO solution
  • Reduces reagent costs
  • Eliminates the need for intermediate dilutions
  • Provides excellent mixing
  • Length: 53mm
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.
Description & Specifications
Specifications
10 x 96/Rack