Thermo Scientific™ Reagents for EpiJET™ Bisulfite Conversion Kit
Perform on-column desulfonation of converted DNA that has been bound to the membrane of a micro column. Thermo Scientific™ Desulfonation Buffer for EpiJET™ Bisulfite Conversion Kit provides additional reagent for use in bisulfite conversion of DNA for methylation analysis.
$52.74 - $63.74
Specifications
For Use With (Equipment) EpiJET Bisulfite Conversion Kit
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
FERR2131
View Documents
Thermo Scientific™
R2131
25mL
Each for $52.74
FERR2151
View Documents
Thermo Scientific™
R2151
3.5mL
Each for $63.74
Description
Description
The EpiJET Bisulfite Conversion Kit is designed for simple and reliable bisulfite conversion of DNA for methylation analysis. The kit integrates the thermal DNA denaturation and the bisulfite conversion reactions into one step. The unmethylated cytosines are deaminated and converted to uracils using powdered Modification Reagent that has been reconstituted with the two Modification Solutions. This step can be performed following long or short protocol, depending on the downstream application. The converted DNA is bound to a membrane for on-column desulfonation and subsequent purification steps, and then eluted. The resulting elution of the converted DNA is suitable for a number of techniques used in methylation status analysis, including PCR, qPCR, COBRA and sequencing.
Specifications
Specifications
EpiJET Bisulfite Conversion Kit
SDS