Please login to your online account to display your discounted pricing

Thermo Scientific™ Phire™ Tissue Direct PCR Master Mix


Amplify DNA directly from variety of human and animal samples with no prior purification with Phire Tissue Direct PCR Master Mix.

Manufacturer: thermo scientific™  F170S

 View more versions of this product

Catalog No. F170S

  • / Each
 

Description & Specifications

Specifications

Storage Requirements Store all components of kit at -20deg.C upon arrival. The Dilution Buffer can also be stored at 4deg.C once it is thawed.
For Use With (Application) Genotyping, Transgene detection, Knockout analysis, Sequencing
Quantity 250 x 20μL Reactions
Concentration 2X
Includes 2X Phire Tissue Direct PCR Master Mix, Dilution Buffer, DNARelease Additive, Universal control primer mix (25μM each), nuclease- free Water, O'GeneRuler Express DNA Ladder