Please login to your online account to display your discounted pricing

Thermo Scientific™ Phire™ Tissue Direct PCR Master Mix


Amplify DNA directly from variety of human and animal samples with no prior purification with Phire Tissue Direct PCR Master Mix.

Manufacturer: thermo scientific™  F170L

 View more versions of this product

Catalog No. F170L

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Genotyping, Transgene detection, Knockout analysis, Sequencing
Includes 2X Phire Tissue Direct PCR Master Mix, Dilution Buffer, DNARelease Additive, Universal control primer mix (25μM each), nuclease- free Water, O'GeneRuler Express DNA Ladder
Quantity 1250 x 20μL Reactions
Storage Requirements Store all components of kit at -20deg.C upon arrival. The Dilution Buffer can also be stored at 4deg.C once it is thawed.
Concentration 2X
Concentration 2X