Please login to your online account to display your discounted pricing

Thermo Scientific™ MagJET™ Whole Blood gDNA Kit


Purifies genomic DNA from fresh or frozen whole blood treated with EDTA or citrate, using magnetic bead technology.

Manufacturer: thermo scientific™  K2741

 View more versions of this product

Catalog No. FERK2741

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) PCR and qPCR, FastDigest and conventional restriction digestion, Southern Blotting
Includes Lysis Buffer (15mL), MagJET Magnetic Beads (2 x 1.4mL), Proteinase K (2 x 1mL), Binding Buffer (23mL), conc. Wash Buffer 1 (100mL), conc. Wash Buffer 2 (40mL), Elution Buffer (30mL)
Shelf Life 3 Years
Quantity 96 Preparations
Storage Requirements Store Proteinase K solution at room temperature as long as the vial remains sealed. After opening, store at -20deg.C. MagJET Magnetic Beads should be stored at 4deg.C. Other components of the kit should be stored at room temperature (15deg. to 25deg.C).
Quantity 96 Preparations