Please login to your online account to display your discounted pricing

Thermo Scientific™ MagJET™ Viral DNA and RNA Kit


Purifies viral nucleic acids from human and animal samples, including plasma, serum, saliva, and urine, using magnetic bead technology amenable to high-throughput operations.

Manufacturer: thermo scientific™  K2781

 View more versions of this product

Catalog No. FERK2781

  • / Each
 

Description & Specifications

Specifications

Shelf Life 3 Years
Quantity 96 Preparations
Storage Requirements Store Carrier RNA in the original aluminum bag at -20deg.C. Once open, store Proteinase K solution at -20deg.C. MagJET Magnetic Beads should be stored at 4deg.C. Other components of the kit should be stored at room temperature (15deg. to 25deg.C).
For Use With (Application) Downstream use in qualitative and quantitative nucleic acid based assays like RT-PCR, RT-qPCR
Includes Lysis Buffer, MagJET Magnetic Beads, Proteinase K, Carrier RNA, Wash Buffer 1 (conc.), Wash Buffer 2 (conc.), nuclease free water
Accessories

3 Years