Thermo Scientific™ MagJET™ Separation Rack
The MagJET separation rack allows fast and efficient procedures for magnetic bead-based nucleic acid isolation and affinity purification workflows.
$125.00 - $448.00
Specifications
Products ${productFamilyLength}
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
FERMR02 Thermo Scientific™
MR02
Each for $322.00
FERMR01 Thermo Scientific™
MR01
Each for $125.00
FERMR04 Thermo Scientific™
MR04
Each for $284.00
FERMR03 Thermo Scientific™
MR03
Each for $448.00
Specifications
Description
MagJET Separation Racks are magnetic separation devices intended for low- to high- throughput magnetic bead manipulation procedures such as magnetic bead-based nucleic acid isolation. MagJET Separation Racks contain high-performance permanent neodymium magnets embedded in a plastic separation unit frame. In combination with MagJET Magnetic Beads MagJET Separation Racks allow fast and efficient magnetic bead-based separation procedures.
MagJET Separation Rack is a magnetic separation device intended for low-throughput magnetic bead manipulation procedures such as magnetic bead-based nucleic acid isolation. The MagJET Separation Rack contains a high-performance permanent neodymium magnet embedded in a plastic separation unit frame. In combination with MagJET Magnetic Beads the MagJET Separation Rack allows fast and efficient magnetic bead-based separation procedures.
Highlights
• Contains a powerful permanent neodymium magnet for instantaneous bead collection from the solution
• Specially designed for robust performance with different sample volumes
• Stable and tight magnetic bead capture on microcentrifuge tube wall
• Highly efficient nucleic acid purification in combination with Thermo Scientific MagJET magnetic-bead based kits Applications
• Affinity purification using magnetic particles. Includes
• MagJET Separation Rack unit
Description & Specifications
Specifications
0.0