Please login to your online account to display your discounted pricing

Thermo Scientific™ Luminaris Probe Low ROX qPCR Master Mixes


Ready-to-use qPCR master mixes with premixed UDG optimized for probe detection using standard cycling protocol. Master mixes available with or without blue color.

Manufacturer: thermo scientific™  K0942

 View more versions of this product

Catalog No. FERK0942

  • / Each
 

Description & Specifications

Specifications

Includes 2x Luminaris Probe Low ROX qPCR Master Mix (4 X 1.25mL): includes Hot Start Taq Polymerase, UDG and dNTPs (also dUTP) in an optimized colorless PCR buffer; supplied with 40x Yellow Sample Buffer and (4 x 1.25mL) nuclease-free water
Storage Requirements Store the master mix at -20deg.C for long term storage or at 4deg.C for up to one month. ROX dye is sensitive to light and should be protected from direct light. When stored at -20deg.C, full activity of the mix is retained for at least 24 months as indicated on the tube label.
Shelf Life 2 Years
For Use With (Application) Gene expression analysis, siRNA validation, Genotyping, Pathogen detection
Concentration 2X
Quantity 500 x 20μL reactions
Accessories

2 Years