Please login to your online account to display your discounted pricing

Thermo Scientific™ Luminaris™ Color HRM Master Mix


Ready-to-use master mix for easy and accurate discrimination of DNA sequence variants by High-Resolution Melting (HRM) analysis.

Manufacturer: thermo scientific™  K1032

 View more versions of this product

Catalog No. FERK1032

  • / Each
 

Description & Specifications

Specifications

Quantity 1250 x 20μL reactions
Includes Luminaris Color HRM Master Mix (2X) includes chemically modified Hot Start Taq DNA Polymerase, EvaGreen™ fluorescent dye, dNTPs and MgCl2 in an optimized PCR buffer with a blue dye; 40x Yellow Sample Buffer with a yellow dye; Nuclease-free water
Concentration 2X
Shelf Life 2 Years
For Use With (Application) SNP genotyping, Mutation discovery (gene scanning), Heterozygosity screening, Bacterial genotyping, CpG methylation analysis
Storage Requirements Store at -20deg.C in dark for long term storage or at 4deg.C for up to three weeks. When stored at -20deg.C, full activity of the mix is retained for at least 24 months as indicated on the tube label. Repeated freezing and thawing of the Master Mix is not recommended.