Please login to your online account to display your discounted pricing

Thermo Scientific™ ClaSeek™ Library Preparation Kit, Ion Torrent™ Compatible


Generate Illumina™-compatible next-gen sequencing libraries from low sample input using a one-tube PCR-free method with optimized master mixes

Manufacturer: thermo scientific™  K1352

 View more versions of this product

Catalog No. FERK1352

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) DNA fragment library preparation for downstream use in next generation sequencing workflow.
Storage Requirements Store at -20deg.C. Cleanup Columns must be stored at room temperature.
Shelf Life 1 Year
Includes ClaSeek End Conversion Master Mix IT, ClaSeek Ligation Mix IT, Adapters IT, Library Amplification Primer Mix, nuclease-free water, Cleanup Columns (pre-assembled with collection tubes)
Quantity 50 reactions
Accessories

1 Year