Please login to your online account to display your discounted pricing

Thermo Scientific™ ClaSeek™ Library Preparation Kit, Illumina™ Compatible


Generate Illumina™-compatible next-gen sequencing libraries from low sample input using a one-tube PCR-free method with optimized master mixes

Manufacturer: thermo scientific™  K1341

 View more versions of this product

Catalog No. FERK1341

  • / Each
 

Description & Specifications

Specifications

Storage Requirements Store at -20deg.C before opening.
Shelf Life 1 Year
Includes ClaSeek End Conversion Master Mix IL, ClaSeek Ligation Mix IL, nuclease-free water
For Use With (Application) DNA fragment library preparation for downstream use in next generation sequencing workflow.
Quantity 12 reactions
Accessories

1 Year