Please login to your online account to display your discounted pricing
Thermo Scientific™ F-G1 Fixed Angle Adapters For CL10™ Rotor
For Thermo Scientific™ IEC F-G1 Fixed Angle rotor
Manufacturer: thermo scientific™  11210542
 View more versions of this product
Catalog No. 05-376-241
  • / Pack of 6 Each
Description & Specifications
Specifications
Type Adapter
Product Type Adapter
Capacity (Metric) 1 x 2/1.5mL
Tube Size 10mm
For Use With (Equipment) Thermo Scientific™ IEC™ F-G1 fixed angle rotor
Angle fixed-angle