Please login to your online account to display your discounted pricing
Thermo Scientific™ Evolution 300 and 600 Spectrophotometer Smart Sipper System
Easy-to-use sampling accessory featuring a reliable multi-roller peristaltic pump
$81.58 - $81.58
Specifications
For Use With (Equipment) Evolution™ 300 and 600 spectrophotometer Smart Sipper System
View More Specs
Requires:  Silicone pump tubing (14-386-424) or Acid pump tubing (14-386-425) and flow cell
Products
Catalog Number Mfr. No. Price Quantity    
14-386-424
thermo scientific™
401322844561
Each for $81.58
Description & Specifications
Specifications
For Use With (Equipment) Evolution™ 300 and 600 spectrophotometer Smart Sipper System
  • For use with Thermo Scientific™ Evolution™ 300 and 600 Spectrophotometers
  • Complete and automated integration into VISION and Local Control software methods
  • Automatic recognition by software, smart alignment and serial number reporting
  • Handles sample volumes from 200μL to 9.999mL with customizable sip volume and air gap
  • Methods can be automated for sample measurement after they are sipped