missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Dri-Bath™ Incubator Modular Blocks

For use with Thermo Scientific Dri-Bath Incubators

Manufacturer:  Thermo Scientific™ BK165X5AQ

 View more versions of this product

Catalog No. 1171656Q



Description

Description

Modular Blocks

  • For use with Thermo Scientific™ Dri-Bath™ Modular Incubators
  • Anodized aluminum alloy
  • Each hole 48mm deep
  • Thermometer holes

Specifications

Specifications

Accessory, Block
4 in.
4.8 cm
Dri-bath incubator
5 cm
3 in.
1.8 in.
10.1 cm
Aluminum Alloy
2 in.
4.5 mL
7.6 cm
SDS
Documents